Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACTTGAAAGTAATAAAAATGTT[A/C]TTTAAAAGTATTGTATGATACAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603907 MIM: 603930 MIM: 608007 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2S1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2S1 - eukaryotic translation initiation factor 2 subunit alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004094.4 | 4155 | UTR 3 | NP_004085.1 |
PLEK2 - pleckstrin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016445.2 | 4155 | Intron | NP_057529.1 | |||
XM_011536644.2 | 4155 | Intron | XP_011534946.1 |