Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAATTCTCTTCCTTAAAAAAC[A/T]GCCATTTCAAAAGATTTTCTTTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 102575 | ||||||||||||||||||||
Literature Links: |
ACTN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTN1 - actinin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102.3 | 4175 | UTR 3 | NP_001093.1 | |||
NM_001130004.1 | 4175 | UTR 3 | NP_001123476.1 | |||
NM_001130005.1 | 4175 | UTR 3 | NP_001123477.1 | |||
XM_011537265.2 | 4175 | UTR 3 | XP_011535567.1 | |||
XM_011537266.2 | 4175 | UTR 3 | XP_011535568.1 | |||
XM_011537267.2 | 4175 | UTR 3 | XP_011535569.1 | |||
XM_011537268.2 | 4175 | UTR 3 | XP_011535570.1 | |||
XM_017021720.1 | 4175 | UTR 3 | XP_016877209.1 | |||
XM_017021721.1 | 4175 | UTR 3 | XP_016877210.1 | |||
XM_017021722.1 | 4175 | UTR 3 | XP_016877211.1 | |||
XM_017021723.1 | 4175 | UTR 3 | XP_016877212.1 | |||
XM_017021725.1 | 4175 | UTR 3 | XP_016877214.1 | |||
XM_017021726.1 | 4175 | UTR 3 | XP_016877215.1 | |||
XM_017021727.1 | 4175 | UTR 3 | XP_016877216.1 | |||
XM_017021728.1 | 4175 | UTR 3 | XP_016877217.1 |