Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCTGAAAGTACTTTCAAAGGAG[A/G]CCTGAAGTAGAAAGGAAAGGTATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ERH - enhancer of rudimentary homolog (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A9 - solute carrier family 39 member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252148.1 | Intron | NP_001239077.1 | ||||
NM_001252150.1 | Intron | NP_001239079.1 | ||||
NM_001252151.1 | Intron | NP_001239080.1 | ||||
NM_001252152.1 | Intron | NP_001239081.1 | ||||
NM_018375.4 | Intron | NP_060845.2 | ||||
XM_005267834.1 | Intron | XP_005267891.1 | ||||
XM_017021426.1 | Intron | XP_016876915.1 |