Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTCACACAAGGGAAGGAGGAGG[A/G]AGGGAGGTCTCCAATGTGGGGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K9 - mitogen-activated protein kinase kinase kinase 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284230.1 | 7513 | UTR 3 | NP_001271159.1 | |||
NM_001284231.1 | 7513 | UTR 3 | NP_001271160.1 | |||
NM_001284232.1 | 7513 | UTR 3 | NP_001271161.1 | |||
NM_033141.3 | 7513 | UTR 3 | NP_149132.2 | |||
XM_005267683.4 | 7513 | UTR 3 | XP_005267740.1 | |||
XM_011536788.2 | 7513 | UTR 3 | XP_011535090.1 | |||
XM_011536792.1 | 7513 | UTR 3 | XP_011535094.1 | |||
XM_011536794.2 | 7513 | UTR 3 | XP_011535096.1 |