Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGCCACAGCCAGAGACCGAGGA[A/G]GAGGACTTCAAAGAGGGGGAGCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
SAMD15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SAMD15 - sterile alpha motif domain containing 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010860.1 | 183 | Silent Mutation | GAA,GAG | E,E 61 | NP_001010860.1 |
TMED8 - transmembrane p24 trafficking protein family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_213601.1 | 183 | Intron | NP_998766.1 | |||
XM_017021223.1 | 183 | Intron | XP_016876712.1 | |||
XM_017021224.1 | 183 | Intron | XP_016876713.1 | |||
XM_017021225.1 | 183 | Intron | XP_016876714.1 |