Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTATCAGGTCGGGTGTGTTGGAAC[C/T]CCAGGTTGCCGAGCAGCCGTGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608466 MIM: 613401 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHSA1 - activator of Hsp90 ATPase activity 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321441.1 | Intron | NP_001308370.1 | ||||
NM_012111.2 | Intron | NP_036243.1 |
VIPAS39 - VPS33B interacting protein, apical-basolateral polarity regulator, spe-39 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |