Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTCCCCAGACAGCATGGAAC[A/G]CAGGCAGAAGCGCGCCGTGGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608466 MIM: 612684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHSA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AHSA1 - activator of Hsp90 ATPase activity 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ISM2 - isthmin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182509.3 | 1393 | UTR 3 | NP_872315.2 | |||
NM_199296.2 | 1393 | Missense Mutation | CGT,TGT | R,C 477 | NP_954993.1 | |
XM_011536489.1 | 1393 | Missense Mutation | CGT,TGT | R,C 389 | XP_011534791.1 |