Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTACTGGCATCTCATGAAGTAT[C/T]AAAAATCAACAGCTTTTAGTGAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607958 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STXBP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STXBP6 - syntaxin binding protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304476.1 | Intron | NP_001291405.1 | ||||
NM_001304477.1 | Intron | NP_001291406.1 | ||||
NM_014178.7 | Intron | NP_054897.4 | ||||
XM_006720121.3 | Intron | XP_006720184.1 | ||||
XM_011536669.2 | Intron | XP_011534971.1 | ||||
XM_017021231.1 | Intron | XP_016876720.1 | ||||
XM_017021232.1 | Intron | XP_016876721.1 | ||||
XM_017021233.1 | Intron | XP_016876722.1 | ||||
XM_017021234.1 | Intron | XP_016876723.1 | ||||
XM_017021235.1 | Intron | XP_016876724.1 | ||||
XM_017021236.1 | Intron | XP_016876725.1 | ||||
XM_017021237.1 | Intron | XP_016876726.1 | ||||
XM_017021238.1 | Intron | XP_016876727.1 | ||||
XM_017021239.1 | Intron | XP_016876728.1 | ||||
XM_017021240.1 | Intron | XP_016876729.1 | ||||
XM_017021241.1 | Intron | XP_016876730.1 | ||||
XM_017021242.1 | Intron | XP_016876731.1 |