Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCGTTCCACTGGACAGGGAAAG[C/T]TTGAAACTTGGGCTCTGCCGTCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610351 | ||||||||||||||||||||
Literature Links: |
PPP4R3A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PPP4R3A - protein phosphatase 4 regulatory subunit 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284280.1 | 2318 | Silent Mutation | AAA,AAG | K,K 732 | NP_001271209.1 | |
NM_001284281.1 | 2318 | Silent Mutation | AAA,AAG | K,K 506 | NP_001271210.1 | |
XM_005267842.2 | 2318 | Silent Mutation | AAA,AAG | K,K 745 | XP_005267899.1 |