Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGACACAGCACTGATATGGATG[C/T]AGCTCTGCAGTCTTTGCCCTGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00341 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00341 - long intergenic non-protein coding RNA 341 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNE3 - spectrin repeat containing nuclear envelope family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152592.3 | 3042 | UTR 3 | NP_689805.3 | |||
XM_005267377.3 | 3042 | UTR 3 | XP_005267434.1 | |||
XM_006720063.3 | 3042 | UTR 3 | XP_006720126.1 | |||
XM_011536515.2 | 3042 | UTR 3 | XP_011534817.1 | |||
XM_017021056.1 | 3042 | UTR 3 | XP_016876545.1 | |||
XM_017021057.1 | 3042 | Intron | XP_016876546.1 |