Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGACGACCCGGAGCTCCGACA[A/G]GGTCAGGCCAGTGTGCCGGGCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609588 | ||||||||||||||||||||
Literature Links: |
GLRX5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLRX5 - glutaredoxin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016417.2 | 398 | Silent Mutation | CAA,CAG | Q,Q 98 | NP_057501.2 |
SCARNA13 - small Cajal body-specific RNA 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNHG10 - small nucleolar RNA host gene 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |