Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGAATCTCTGTATAGTTTCAA[A/G]TAGAATTAATTTTCAAGCTCAATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605553 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC730202 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC730202 - uncharacterized LOC730202 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAPOLA - poly(A) polymerase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252006.1 | Intron | NP_001238935.1 | ||||
NM_001252007.1 | Intron | NP_001238936.1 | ||||
NM_001293627.1 | Intron | NP_001280556.1 | ||||
NM_001293628.1 | Intron | NP_001280557.1 | ||||
NM_001293632.1 | Intron | NP_001280561.1 | ||||
NM_032632.4 | Intron | NP_116021.2 | ||||
XM_005267281.1 | Intron | XP_005267338.1 | ||||
XM_005267282.2 | Intron | XP_005267339.1 | ||||
XM_017020935.1 | Intron | XP_016876424.1 | ||||
XM_017020936.1 | Intron | XP_016876425.1 |