Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGAAAAGGAACCTCAGAGACCT[G/T]TTTCCACCAAGAGCCCAGACAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603544 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC85C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC85C - coiled-coil domain containing 85C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144995.1 | 2739 | UTR 3 | NP_001138467.1 | |||
XM_011536705.2 | 2739 | UTR 3 | XP_011535007.1 | |||
XM_011536706.2 | 2739 | Intron | XP_011535008.1 | |||
XM_011536707.2 | 2739 | Intron | XP_011535009.1 |
CCNK - cyclin K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |