Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAAGTCATGGGCCCCTTGGAA[C/T]GAGCTTCAGGATGACACTGCACTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604087 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP46A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP46A1 - cytochrome P450 family 46 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006668.1 | Intron | NP_006659.1 | ||||
XM_005267274.4 | Intron | XP_005267331.1 | ||||
XM_011536364.1 | Intron | XP_011534666.1 | ||||
XM_011536365.1 | Intron | XP_011534667.1 | ||||
XM_017020933.1 | Intron | XP_016876422.1 |
HHIPL1 - HHIP like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |