Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTCTTGAGCTCACCGGTGAGCA[G/T]GGCTCCGCTGGTGTAATCCTGCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609911 MIM: 191050 | ||||||||||||||||||||
Literature Links: |
SLC25A47 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC25A47 - solute carrier family 25 member 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WARS - tryptophanyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004184.3 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | NP_004175.2 | |
NM_173701.1 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | NP_776049.1 | |
NM_213645.1 | 1628 | Missense Mutation | ATG,CTG | M,L 384 | NP_998810.1 | |
NM_213646.1 | 1628 | Missense Mutation | ATG,CTG | M,L 384 | NP_998811.1 | |
XM_005268044.3 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_005268101.1 | |
XM_006720249.3 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_006720312.1 | |
XM_011537133.2 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_011535435.1 | |
XM_011537135.2 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_011535437.1 | |
XM_011537136.2 | 1628 | Missense Mutation | ATG,CTG | M,L 384 | XP_011535438.1 | |
XM_017021627.1 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_016877116.1 | |
XM_017021628.1 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_016877117.1 | |
XM_017021629.1 | 1628 | Missense Mutation | ATG,CTG | M,L 425 | XP_016877118.1 |