Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCCCCTCGCTGGGTGCATAGCC[A/G]GGCAGTGGGTCAGCCGAGCGGCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BEGAIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BEGAIN - brain enriched guanylate kinase associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159531.1 | 1821 | Silent Mutation | CCC,CCT | P,P 511 | NP_001153003.1 | |
NM_020836.3 | 1821 | Silent Mutation | CCC,CCT | P,P 511 | NP_065887.1 | |
XM_005267919.3 | 1821 | Silent Mutation | CCC,CCT | P,P 524 | XP_005267976.1 | |
XM_005267920.4 | 1821 | Silent Mutation | CCC,CCT | P,P 523 | XP_005267977.1 | |
XM_005267921.3 | 1821 | Silent Mutation | CCC,CCT | P,P 511 | XP_005267978.1 | |
XM_011537031.1 | 1821 | Silent Mutation | CCC,CCT | P,P 530 | XP_011535333.1 | |
XM_017021521.1 | 1821 | Silent Mutation | CCC,CCT | P,P 511 | XP_016877010.1 | |
XM_017021522.1 | 1821 | Silent Mutation | CCC,CCT | P,P 502 | XP_016877011.1 |
WDR25 - WD repeat domain 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |