Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAACTGGGGGAAGTGGGGCCTG[C/T]GCTCGCTGTCCACACACTCCATGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164730 | ||||||||||||||||||||
Literature Links: |
AKT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AKT1 - AKT serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014431.1 | 1734 | Missense Mutation | CAC,CGC | H,R 465 | NP_001014431.1 | |
NM_001014432.1 | 1734 | Missense Mutation | CAC,CGC | H,R 465 | NP_001014432.1 | |
NM_005163.2 | 1734 | Missense Mutation | CAC,CGC | H,R 465 | NP_005154.2 | |
XM_005267401.1 | 1734 | Missense Mutation | CAC,CGC | H,R 465 | XP_005267458.1 | |
XM_017021075.1 | 1734 | Intron | XP_016876564.1 | |||
XM_017021076.1 | 1734 | Intron | XP_016876565.1 | |||
XM_017021077.1 | 1734 | Intron | XP_016876566.1 | |||
XM_017021078.1 | 1734 | Intron | XP_016876567.1 |