Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAAATCTTCATTATCCATGTC[A/G]ATTTCTTTCCAATGCTTATCAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBGCP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBGCP3 - tubulin gamma complex associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286277.1 | 4459 | UTR 3 | NP_001273206.1 | |||
NM_001286278.1 | 4459 | Intron | NP_001273207.1 | |||
NM_001286279.1 | 4459 | Intron | NP_001273208.1 | |||
NM_006322.5 | 4459 | UTR 3 | NP_006313.1 | |||
XM_005268293.1 | 4459 | Intron | XP_005268350.1 | |||
XM_011537462.2 | 4459 | UTR 3 | XP_011535764.1 | |||
XM_011537463.2 | 4459 | UTR 3 | XP_011535765.1 | |||
XM_011537464.2 | 4459 | UTR 3 | XP_011535766.1 | |||
XM_017020323.1 | 4459 | UTR 3 | XP_016875812.1 |