Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACCAGAATACAGTTTGATTTTTT[A/T]GGAGCTTTTATTGTTTTAGTAATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182120 | ||||||||||||||||||||
Literature Links: |
SPARC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPARC - secreted protein acidic and cysteine rich | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001309443.1 | 3228 | UTR 3 | NP_001296372.1 | |||
NM_001309444.1 | 3228 | UTR 3 | NP_001296373.1 | |||
NM_003118.3 | 3228 | UTR 3 | NP_003109.1 |