Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCTGACGCCCGGAGCCCGCTTTA[C/T]TCTCCCGAATGCCCAAGATGACGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606307 MIM: 606316 MIM: 606317 MIM: 606318 MIM: 606319 MIM: 606308 MIM: 606309 MIM: 606310 MIM: 606311 MIM: 606312 MIM: 606313 MIM: 606314 MIM: 606315 MIM: 606320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDHA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCDHA1 - protocadherin alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018900.3 | 607 | Intron | NP_061723.1 | |||
NM_031410.2 | 607 | Intron | NP_113598.1 | |||
NM_031411.2 | 607 | Intron | NP_113599.1 |
PCDHA10 - protocadherin alpha 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018901.3 | 607 | Intron | NP_061724.1 | |||
NM_031859.2 | 607 | Intron | NP_114065.1 | |||
NM_031860.2 | 607 | Intron | NP_114066.1 |
PCDHA11 - protocadherin alpha 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018902.4 | 607 | Intron | NP_061725.1 | |||
NM_031861.2 | 607 | Intron | NP_114067.1 |
PCDHA12 - protocadherin alpha 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018903.3 | 607 | Intron | NP_061726.1 | |||
NM_031864.2 | 607 | Intron | NP_114070.1 |
PCDHA13 - protocadherin alpha 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018904.2 | 607 | Intron | NP_061727.1 | |||
NM_031865.1 | 607 | Intron | NP_114071.1 |
PCDHA2 - protocadherin alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018905.2 | 607 | Intron | NP_061728.1 | |||
NM_031495.1 | 607 | Intron | NP_113683.1 | |||
NM_031496.1 | 607 | Intron | NP_113684.1 |
PCDHA3 - protocadherin alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018906.2 | 607 | Intron | NP_061729.1 | |||
NM_031497.1 | 607 | Intron | NP_113685.1 |
PCDHA4 - protocadherin alpha 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018907.3 | 607 | Intron | NP_061730.1 | |||
NM_031500.2 | 607 | Intron | NP_113688.1 |
PCDHA5 - protocadherin alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018908.2 | 607 | Intron | NP_061731.1 | |||
NM_031501.1 | 607 | Intron | NP_113689.1 |
PCDHA6 - protocadherin alpha 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018909.3 | 607 | Intron | NP_061732.1 | |||
NM_031848.2 | 607 | Intron | NP_114036.1 | |||
NM_031849.2 | 607 | Intron | NP_114037.1 |
PCDHA7 - protocadherin alpha 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018910.2 | 607 | Intron | NP_061733.1 | |||
NM_031852.1 | 607 | Intron | NP_114040.1 |
PCDHA8 - protocadherin alpha 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018911.2 | 607 | Intron | NP_061734.1 | |||
NM_031856.1 | 607 | Intron | NP_114062.1 |
PCDHA9 - protocadherin alpha 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014005.4 | 607 | Intron | NP_054724.1 | |||
NM_031857.1 | 607 | Intron | NP_114063.1 |
PCDHAC1 - protocadherin alpha subfamily C, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018898.3 | 607 | Missense Mutation | ACT,ATT | T,I 144 | NP_061721.2 | |
NM_031882.3 | 607 | Missense Mutation | ACT,ATT | T,I 144 | NP_114088.2 |