Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCCAATGTGCATGAGTTTAATG[A/G]ACCAGATAGATTATTGTATCGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151350 | ||||||||||||||||||||
Literature Links: |
LARS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LARS - leucyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317964.1 | 3738 | Silent Mutation | NP_001304893.1 | |||
NM_001317965.1 | 3738 | Silent Mutation | NP_001304894.1 | |||
NM_016460.3 | 3738 | Silent Mutation | NP_057544.2 | |||
NM_020117.10 | 3738 | Silent Mutation | NP_064502.9 | |||
XM_011537656.2 | 3738 | Silent Mutation | XP_011535958.1 |
PLAC8L1 - PLAC8 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |