Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCCAGGAGTCTGGACAGGAGGC[C/T]AGGCGCTGCATGCAGCCCCAGTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606542 MIM: 612187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HDAC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HDAC7 - histone deacetylase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098416.3 | 2916 | Silent Mutation | CTA,CTG | L,L 903 | NP_001091886.1 | |
NM_001308090.1 | 2916 | Silent Mutation | CTA,CTG | L,L 923 | NP_001295019.1 | |
NM_015401.4 | 2916 | Silent Mutation | CTA,CTG | L,L 940 | NP_056216.2 | |
XM_011538478.1 | 2916 | Silent Mutation | CTA,CTG | L,L 984 | XP_011536780.1 | |
XM_011538479.1 | 2916 | Silent Mutation | CTA,CTG | L,L 947 | XP_011536781.1 | |
XM_011538480.1 | 2916 | Silent Mutation | CTA,CTG | L,L 947 | XP_011536782.1 | |
XM_011538481.1 | 2916 | Silent Mutation | CTA,CTG | L,L 901 | XP_011536783.1 | |
XM_011538482.1 | 2916 | Silent Mutation | CTA,CTG | L,L 901 | XP_011536784.1 | |
XM_011538483.1 | 2916 | Intron | XP_011536785.1 | |||
XM_017019455.1 | 2916 | Silent Mutation | CTA,CTG | L,L 967 | XP_016874944.1 | |
XM_017019456.1 | 2916 | Intron | XP_016874945.1 |
SLC48A1 - solute carrier family 48 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |