Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGATCAAAGAATATCTCATTAAA[C/T]GATCTCTGGCTAGGTAGTTTTCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 143055 MIM: 615488 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCNT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCNT1 - cyclin T1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001240.3 | 3898 | UTR 3 | NP_001231.2 | |||
NM_001277842.1 | 3898 | UTR 3 | NP_001264771.1 | |||
XM_017020197.1 | 3898 | UTR 3 | XP_016875686.1 |
KANSL2 - KAT8 regulatory NSL complex subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |