Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATTAACACACAGTGGAGAACTG[C/T]CCTTCGGGAAGTCAAGACCAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611088 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC65 - coiled-coil domain containing 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286957.1 | 430 | UTR 5 | NP_001273886.1 | |||
NM_033124.4 | 430 | Missense Mutation | GCC,GTC | A,V 68 | NP_149115.2 |