Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGACAGATGGTGGAAATCCCC[A/G]TTTTCCATAGGCCAAGTGAGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103190 MIM: 611088 MIM: 610571 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARF3 - ADP ribosylation factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCDC65 - coiled-coil domain containing 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FKBP11 - FK506 binding protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143781.1 | 277 | Missense Mutation | CGG,TGG | R,W 20 | NP_001137253.1 | |
NM_001143782.1 | 277 | Missense Mutation | CGG,TGG | R,W 122 | NP_001137254.1 | |
NM_016594.2 | 277 | Missense Mutation | CGG,TGG | R,W 122 | NP_057678.1 |