Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGTTTCTGGTTCTTTTTTTATTTA[A/G]CAGAAATCTTTGCTTTTTTCTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602113 MIM: 602742 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMT2D - lysine methyltransferase 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003482.3 | Intron | NP_003473.3 | ||||
XM_005269162.4 | Intron | XP_005269219.1 | ||||
XM_006719614.3 | Intron | XP_006719677.1 | ||||
XM_006719616.3 | Intron | XP_006719679.1 | ||||
XM_011538770.2 | Intron | XP_011537072.1 | ||||
XM_011538771.2 | Intron | XP_011537073.1 | ||||
XM_011538772.2 | Intron | XP_011537074.1 | ||||
XM_011538773.2 | Intron | XP_011537075.1 | ||||
XM_011538774.2 | Intron | XP_011537076.1 | ||||
XM_011538776.2 | Intron | XP_011537078.1 |
PRKAG1 - protein kinase AMP-activated non-catalytic subunit gamma 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |