Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACACTTAGGAGAAGGTTCCCCTC[C/T]TTTACTTCTGTTTCTCTGCTGATAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBA1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBA1C - tubulin alpha 1c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303114.1 | Intron | NP_001290043.1 | ||||
NM_001303115.1 | Intron | NP_001290044.1 | ||||
NM_001303116.1 | Intron | NP_001290045.1 | ||||
NM_001303117.1 | Intron | NP_001290046.1 | ||||
NM_032704.4 | Intron | NP_116093.1 |