Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATGGAACCGTAAGCACTCAAAG[C/T]ATGGTCTACCTCTGCTTGCCTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCDIN3D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCDIN3D - BCDIN3 domain containing RNA methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181708.2 | 1615 | UTR 3 | NP_859059.1 |
BCDIN3D-AS1 - BCDIN3D antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCKAP5L - NCK associated protein 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |