Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGATTTTTGCAGCATCCGCTGTG[A/G]GCTGGCTTGGGCTGTGGGCAGCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611159 MIM: 611160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT78 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KRT78 - keratin 78 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300814.1 | 1350 | UTR 3 | NP_001287743.1 | |||
NM_173352.3 | 1350 | UTR 3 | NP_775487.2 | |||
XM_011538010.1 | 1350 | UTR 3 | XP_011536312.1 |
KRT79 - keratin 79 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |