Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCCCTGGCCCCTCTGTAGTGTAG[A/G]AGGGTCCCTGCCCCAATGGCTGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147559 MIM: 180190 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGB7 - integrin subunit beta 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RARG - retinoic acid receptor gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000966.5 | 2239 | UTR 3 | NP_000957.1 | |||
NM_001042728.2 | 2239 | UTR 3 | NP_001036193.1 | |||
NM_001243730.1 | 2239 | UTR 3 | NP_001230659.1 | |||
NM_001243731.1 | 2239 | UTR 3 | NP_001230660.1 | |||
NM_001243732.1 | 2239 | UTR 3 | NP_001230661.1 |