Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGAGCCAGGATCCCCAGCTCC[A/G]CCTGTGGCCCCCCGGCCCCGGGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600447 MIM: 601210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP3K12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP3K12 - mitogen-activated protein kinase kinase kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193511.1 | 2179 | Silent Mutation | GGC,GGT | G,G 660 | NP_001180440.1 | |
NM_006301.3 | 2179 | Silent Mutation | GGC,GGT | G,G 627 | NP_006292.3 | |
XM_005269138.3 | 2179 | Silent Mutation | GGC,GGT | G,G 660 | XP_005269195.1 | |
XM_005269140.2 | 2179 | Intron | XP_005269197.1 | |||
XM_006719588.3 | 2179 | Silent Mutation | GGC,GGT | G,G 660 | XP_006719651.1 | |
XM_011538725.2 | 2179 | Silent Mutation | GGC,GGT | G,G 660 | XP_011537027.1 | |
XM_017019956.1 | 2179 | Silent Mutation | GGC,GGT | G,G 627 | XP_016875445.1 |
PCBP2 - poly(rC) binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |