Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCCTGTCTTCTTTCCCTCTGAC[A/C]AGTACTATGACGACACCTACCCCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615472 MIM: 613787 | ||||||||||||||||||||
Literature Links: |
COPZ1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COPZ1 - coatomer protein complex subunit zeta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271734.1 | Intron | NP_001258663.1 | ||||
NM_001271735.1 | Intron | NP_001258664.1 | ||||
NM_001271736.1 | Intron | NP_001258665.1 | ||||
NM_016057.2 | Intron | NP_057141.1 | ||||
XM_017018997.1 | Intron | XP_016874486.1 |
MIR148B - microRNA 148b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |