Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCAGGCACCCAACAAGGCGGT[A/G]AGTCCTGTGGCCACAATGGTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612104 MIM: 608730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NABP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NABP2 - nucleic acid binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024068.3 | Intron | NP_076973.1 | ||||
XM_005269146.2 | Intron | XP_005269203.1 | ||||
XM_005269147.2 | Intron | XP_005269204.1 | ||||
XM_005269148.4 | Intron | XP_005269205.1 | ||||
XM_005269149.4 | Intron | XP_005269206.1 |
RNF41 - ring finger protein 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A5 - solute carrier family 39 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |