Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTGTCATAGATTTACATCCTTTC[A/C]ATTTTTTGATAATGTCAGACTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602387 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBMS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBMS2 - RNA binding motif single stranded interacting protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002898.3 | Intron | NP_002889.1 | ||||
XM_005269059.4 | Intron | XP_005269116.1 | ||||
XM_005269060.4 | Intron | XP_005269117.1 | ||||
XM_005269061.3 | Intron | XP_005269118.1 | ||||
XM_005269066.4 | Intron | XP_005269123.1 | ||||
XM_006719541.3 | Intron | XP_006719604.1 | ||||
XM_006719542.3 | Intron | XP_006719605.1 | ||||
XM_006719543.3 | Intron | XP_006719606.1 | ||||
XM_006719544.3 | Intron | XP_006719607.1 | ||||
XM_011538637.2 | Intron | XP_011536939.1 | ||||
XM_011538638.2 | Intron | XP_011536940.1 | ||||
XM_011538639.2 | Intron | XP_011536941.1 | ||||
XM_011538640.2 | Intron | XP_011536942.1 | ||||
XM_011538642.2 | Intron | XP_011536944.1 | ||||
XM_017019775.1 | Intron | XP_016875264.1 | ||||
XM_017019776.1 | Intron | XP_016875265.1 | ||||
XM_017019777.1 | Intron | XP_016875266.1 | ||||
XM_017019778.1 | Intron | XP_016875267.1 |