Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTATACCCATTACTGCAAGATCC[C/T]GCCTACCTCCCTGGTGAGAACTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610576 MIM: 156560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP9 - Rho GTPase activating protein 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MARS - methionyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004990.3 | 615 | Silent Mutation | CCC,CCT | P,P 159 | NP_004981.2 | |
XM_006719398.3 | 615 | Intron | XP_006719461.1 |