Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGCCTTTAGACATCCTAGTGG[C/T]GATGTCCAGTTGACAGTTGGCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609677 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OS9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OS9 - OS9, endoplasmic reticulum lectin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017956.2 | Intron | NP_001017956.1 | ||||
NM_001017957.2 | Intron | NP_001017957.1 | ||||
NM_001017958.2 | Intron | NP_001017958.1 | ||||
NM_001261420.1 | Intron | NP_001248349.1 | ||||
NM_001261421.1 | Intron | NP_001248350.1 | ||||
NM_001261422.1 | Intron | NP_001248351.1 | ||||
NM_001261423.1 | Intron | NP_001248352.1 | ||||
NM_006812.3 | Intron | NP_006803.1 | ||||
XM_005268581.1 | Intron | XP_005268638.1 | ||||
XM_006719200.1 | Intron | XP_006719263.1 | ||||
XM_006719201.1 | Intron | XP_006719264.1 |