Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTGATTCGCATGATAGCAGCAGC[A/G]TCTCCCAGCTCTTCTTTTAATTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613397 MIM: 604723 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVIL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AVIL - advillin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006576.3 | 5034 | Silent Mutation | GAC,GAT | D,D 731 | NP_006567.3 | |
XM_017018710.1 | 5034 | Silent Mutation | GAC,GAT | D,D 731 | XP_016874199.1 | |
XM_017018711.1 | 5034 | Silent Mutation | GAC,GAT | D,D 708 | XP_016874200.1 | |
XM_017018712.1 | 5034 | Silent Mutation | GAC,GAT | D,D 708 | XP_016874201.1 | |
XM_017018713.1 | 5034 | Silent Mutation | GAC,GAT | D,D 708 | XP_016874202.1 | |
XM_017018714.1 | 5034 | Silent Mutation | GAC,GAT | D,D 432 | XP_016874203.1 | |
XM_017018715.1 | 5034 | Silent Mutation | GAC,GAT | D,D 414 | XP_016874204.1 |
TSFM - Ts translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172695.1 | 5034 | Intron | NP_001166166.1 | |||
NM_001172696.1 | 5034 | Intron | NP_001166167.1 | |||
NM_001172697.1 | 5034 | Intron | NP_001166168.1 | |||
NM_005726.5 | 5034 | Intron | NP_005717.3 |