Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAGAGAAATGTGTGATGCCTCC[C/T]AGGTTTCTGGCTTGGGAAACTAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616822 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MON2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MON2 - MON2 homolog, regulator of endosome-to-Golgi trafficking | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278469.1 | Intron | NP_001265398.1 | ||||
NM_001278470.1 | Intron | NP_001265399.1 | ||||
NM_001278471.1 | Intron | NP_001265400.1 | ||||
NM_001278472.1 | Intron | NP_001265401.1 | ||||
NM_015026.2 | Intron | NP_055841.2 | ||||
XM_017019041.1 | Intron | XP_016874530.1 | ||||
XM_017019042.1 | Intron | XP_016874531.1 | ||||
XM_017019043.1 | Intron | XP_016874532.1 | ||||
XM_017019044.1 | Intron | XP_016874533.1 | ||||
XM_017019045.1 | Intron | XP_016874534.1 | ||||
XM_017019046.1 | Intron | XP_016874535.1 |