Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTACACAGAATGGTCAGGAAGAGT[C/T]GTTCCTAGACAATGAGATGAGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614539 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HELB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HELB - DNA helicase B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033647.4 | 625 | Missense Mutation | TCG,TTG | S,L 189 | NP_387467.2 | |
XM_005269234.2 | 625 | Missense Mutation | TCG,TTG | S,L 189 | XP_005269291.1 | |
XM_017020230.1 | 625 | Missense Mutation | TCG,TTG | S,L 189 | XP_016875719.1 |