Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCCTTGCACCTGTTAACATCTT[C/T]AAGGCAGGAGCTGATGAAGAGAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605139 MIM: 607743 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT2 - chaperonin containing TCP1 subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198842.1 | 130 | UTR 5 | NP_001185771.1 | |||
NM_006431.2 | 130 | Silent Mutation | TTC,TTT | F,F 12 | NP_006422.1 |
FRS2 - fibroblast growth factor receptor substrate 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-1 - microRNA 3913-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3913-2 - microRNA 3913-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |