Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGCTGAGATGGCTTCTTGCCAGC[C/T]CTCTTTCTACATACATACATACGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615856 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMTC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMTC2 - transmembrane and tetratricopeptide repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320321.1 | Intron | NP_001307250.1 | ||||
NM_001320322.1 | Intron | NP_001307251.1 | ||||
NM_152588.2 | Intron | NP_689801.1 | ||||
XM_017018884.1 | Intron | XP_016874373.1 |