Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACCAGAGAGACACAAACAACCAG[A/T]CCCCATGTTGGATAGCTCAGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC6A15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC6A15 - solute carrier family 6 member 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146335.2 | 2349 | Silent Mutation | GGA,GGT | G,G 511 | NP_001139807.1 | |
NM_018057.6 | 2349 | Intron | NP_060527.2 | |||
NM_182767.5 | 2349 | Silent Mutation | GGA,GGT | G,G 618 | NP_877499.1 | |
XM_011538525.2 | 2349 | Intron | XP_011536827.1 |