Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAATGCTGTGACCAATTATTTG[A/G]GAGTTATAATCTGCAGGCTCCATTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601529 | ||||||||||||||||||||
Literature Links: |
NR2C1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR2C1 - nuclear receptor subfamily 2 group C member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032287.2 | 1743 | Intron | NP_001027458.1 | |||
NM_001127362.1 | 1743 | Intron | NP_001120834.1 | |||
NM_003297.3 | 1743 | Silent Mutation | TCC,TCT | S,S 596 | NP_003288.2 | |
XM_005269133.2 | 1743 | Silent Mutation | TCC,TCT | S,S 591 | XP_005269190.1 | |
XM_006719584.2 | 1743 | Silent Mutation | TCC,TCT | S,S 550 | XP_006719647.1 | |
XM_006719585.2 | 1743 | Silent Mutation | TCC,TCT | S,S 419 | XP_006719648.1 | |
XM_011538716.2 | 1743 | UTR 3 | XP_011537018.1 | |||
XM_011538717.2 | 1743 | Silent Mutation | TCC,TCT | S,S 500 | XP_011537019.1 |