Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCCCATTAAAGGTCCTCAGACTC[C/T]GGGATTTTGGTGGATTTCTTGTGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609457 | ||||||||||||||||||||
Literature Links: |
AMDHD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMDHD1 - amidohydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAL - histidine ammonia-lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258333.1 | 2255 | Silent Mutation | CCA,CCG | P,P 444 | NP_001245262.1 | |
NM_001258334.1 | 2255 | UTR 3 | NP_001245263.1 | |||
NM_002108.3 | 2255 | Silent Mutation | CCA,CCG | P,P 652 | NP_002099.1 | |
XM_011538249.2 | 2255 | Silent Mutation | CCA,CCG | P,P 368 | XP_011536551.1 | |
XM_017019246.1 | 2255 | Silent Mutation | CCA,CCG | P,P 342 | XP_016874735.1 |