Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGACAGTAGTTATTTAGCAGCAA[C/T]GATTCCGCAATAAATAATGCACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK17 - cyclin dependent kinase 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170464.2 | 2517 | UTR 3 | NP_001163935.1 | |||
NM_002595.4 | 2517 | UTR 3 | NP_002586.2 | |||
XM_017019405.1 | 2517 | UTR 3 | XP_016874894.1 | |||
XM_017019406.1 | 2517 | UTR 3 | XP_016874895.1 | |||
XM_017019407.1 | 2517 | UTR 3 | XP_016874896.1 |