Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCAACCTGTTTTAAAGTTTTAT[G/T]TTTACTGTACAAAATAGGATTAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IKBIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IKBIP - IKBKB interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153687.3 | 1416 | Intron | NP_710154.1 | |||
NM_201612.2 | 1416 | UTR 3 | NP_963906.1 | |||
NM_201613.2 | 1416 | UTR 3 | NP_963907.1 |