Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTCACAAATAATACCCTTTCCA[A/G]TAAGGCAGAATCAAGGACCATATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606883 MIM: 610932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IRAK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IRAK4 - interleukin 1 receptor associated kinase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM117 - transmembrane protein 117 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286211.1 | 49 | Intron | NP_001273140.1 | |||
NM_001286212.1 | 49 | Intron | NP_001273141.1 | |||
NM_001286213.1 | 49 | Intron | NP_001273142.1 | |||
NM_032256.2 | 49 | Intron | NP_115632.1 | |||
XM_011538831.2 | 49 | UTR 5 | XP_011537133.1 | |||
XM_011538832.2 | 49 | Intron | XP_011537134.1 | |||
XM_011538833.2 | 49 | Intron | XP_011537135.1 | |||
XM_017020022.1 | 49 | Intron | XP_016875511.1 | |||
XM_017020023.1 | 49 | Intron | XP_016875512.1 |
TWF1 - twinfilin actin binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242397.1 | 49 | Intron | NP_001229326.1 | |||
NM_002822.4 | 49 | Intron | NP_002813.3 |