Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATAAATATGAAAAAGTCTTTGCC[C/T]CTCAGGAGTAATATTGGTTGTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603425 MIM: 612822 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL1 - ADP ribosylation factor like GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177.5 | 2345 | UTR 3 | NP_001168.1 | |||
NM_001301068.1 | 2345 | UTR 3 | NP_001287997.1 |
UTP20 - UTP20, small subunit processome component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |