Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGACGACCTCAGCTCTGGGGAGGC[C/T]GACGTAGACCCAAAGCTCCTGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612986 MIM: 601112 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EID3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EID3 - EP300 interacting inhibitor of differentiation 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008394.2 | 266 | Silent Mutation | GCC,GCT | A,A 21 | NP_001008395.1 |
TXNRD1 - thioredoxin reductase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093771.2 | 266 | Intron | NP_001087240.1 | |||
NM_001261445.1 | 266 | Intron | NP_001248374.1 | |||
NM_001261446.1 | 266 | Intron | NP_001248375.1 | |||
NM_003330.3 | 266 | Intron | NP_003321.3 | |||
NM_182729.2 | 266 | Intron | NP_877393.1 | |||
NM_182742.2 | 266 | Intron | NP_877419.1 | |||
NM_182743.2 | 266 | Intron | NP_877420.1 |