Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCGCGGCCGCCGCAGTCCCGCCG[A/G]CAGCCGCCGCCAACCACCACCATCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTBD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTBD11 - BTB domain containing 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017523.1 | 2752 | Intron | NP_001017523.1 | |||
NM_001018072.1 | 2752 | Missense Mutation | ACA,GCA | T,A 327 | NP_001018082.1 | |
XM_005268645.3 | 2752 | Intron | XP_005268702.1 | |||
XM_011537909.2 | 2752 | Intron | XP_011536211.1 | |||
XM_011537910.2 | 2752 | Intron | XP_011536212.1 | |||
XM_017018807.1 | 2752 | Missense Mutation | ACA,GCA | T,A 327 | XP_016874296.1 |